LIFH309942B6 Life Chemicals Inc.

trans-2-(tert-butylamino)cyclohexan-1-ol

  • MDL number MFCD30495299
  • Molecular Weight : 171.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart