LIFH309942BA Life Chemicals Inc.

(1S,2S)-2-(cyclopentylamino)cyclohexan-1-ol

  • MDL number MFCD20667859
  • Cas Number : 1867063-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart