LIFH309942BD Life Chemicals Inc.

(1R,2R)-2-[cyclobutyl(methyl)amino]cyclopentan-1-ol

  • MDL number MFCD30492491
  • Cas Number : 1932227-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart