LIFH309942C7 Life Chemicals Inc.

2-(chloromethyl)-5-cyclopropyl-1,3,4-thiadiazole

  • MDL number MFCD03789553
  • Cas Number : 138300-59-1
  • Molecular Weight : 174.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart