LIFH309942D4 Life Chemicals Inc.

pent-4-ynal

  • MDL number MFCD11870385
  • Cas Number : 18498-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart