LIFH309942DA Life Chemicals Inc.

(1S,2S)-2-[(prop-2-en-1-yl)amino]cyclopentan-1-ol

  • MDL number MFCD28151492
  • Cas Number : 412035-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart