LIFH309942DC Life Chemicals Inc.

4-[(2,3-dihydro-1H-indol-1-yl)methyl]oxan-4-ol

  • MDL number MFCD28547649
  • Cas Number : 1782382-61-9
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart