LIFH309942DE Life Chemicals Inc.

(1S,2S)-2-(cyclopentylamino)-1-methylcyclobutan-1-ol

  • Cas Number : 2303549-27-9
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart