LIFH309942E2 Life Chemicals Inc.

1-{4-[trans-2-hydroxy-2-methylcyclobutyl]piperazin-1-yl}ethan-1-one

  • Cas Number : 2413365-39-4
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart