LIFH309942E5 Life Chemicals Inc.

(1S,2S)-1-methyl-2-(4-methylpiperidin-1-yl)cyclobutan-1-ol

  • Cas Number : 2303549-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart