LIFH309942E6 Life Chemicals Inc.

(1S,2S)-1-methyl-2-{[(1r,4r)-4-methylcyclohexyl]amino}cyclobutan-1-ol

  • Cas Number : 2303549-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart