LIFH309942F0 Life Chemicals Inc.

4-[(pyrrolidin-1-yl)methyl]oxan-4-ol

  • MDL number MFCD26415301
  • Cas Number : 1565504-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart