LIFH309942FA Life Chemicals Inc.

trans-4-(3,5-dimethylphenoxy)oxolan-3-ol

  • MDL number MFCD31744970
  • Cas Number : 2017938-47-3
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart