LIFH30994305 Life Chemicals Inc.

(1S,2S)-2-(1,4-diazepan-1-yl)-1-methylcyclobutan-1-ol

  • Cas Number : 2303549-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart