LIFH30994308 Life Chemicals Inc.

(1R,2R)-2-[(cyclopropylmethyl)amino]cyclopentan-1-ol

  • MDL number MFCD28151506
  • Cas Number : 1601057-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart