LIFH3099430C Life Chemicals Inc.

ethyl 3-cyclopropyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD08277218
  • Cas Number : 133261-06-0
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart