LIFH3099430F Life Chemicals Inc.

ethyl 2-[(4-methoxyphenyl)sulfanyl]acetate

  • MDL number MFCD05256227
  • Cas Number : 28743-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart