LIFH30994310 Life Chemicals Inc.

4-(2,2,2-trifluoroacetamido)butanoic acid

  • MDL number MFCD00130210
  • Cas Number : 50632-83-2
  • Molecular Weight : 199.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart