LIFH30994317 Life Chemicals Inc.

4-chloro-1-(difluoromethyl)-2-fluorobenzene

  • MDL number MFCD14698547
  • Cas Number : 1214337-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart