LIFH3099431D Life Chemicals Inc.

6-chloro-2-fluoro-3-(trifluoromethyl)benzaldehyde

  • Cas Number : 1026828-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart