LIFH3099431E Life Chemicals Inc.

2-bromo-3-chloro-6-(trifluoromethyl)pyridine-4-carbaldehyde

  • Cas Number : 2303565-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart