LIFH30994321 Life Chemicals Inc.

4-(1-methyl-1H-pyrazol-4-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • Cas Number : 2303565-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart