LIFH30994322 Life Chemicals Inc.

1-(4-ethylphenyl)-3-(1H-indol-3-yl)urea

  • Cas Number : 941987-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart