LIFH30994329 Life Chemicals Inc.

3-bromo-1,5-naphthyridin-2-ol

  • MDL number MFCD00234386
  • Cas Number : 859986-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart