LIFH3099432A Life Chemicals Inc.

3-bromo-1,5-naphthyridin-2-amine

  • Cas Number : 50786-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart