LIFH30994332 Life Chemicals Inc.

2,4-dichloro-3-fluoro-6-(trifluoromethyl)benzaldehyde

  • Cas Number : 2303565-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart