LIFH30994335 Life Chemicals Inc.

3-(difluoromethyl)-2,5,6-trifluorobenzaldehyde

  • Cas Number : 2303565-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart