LIFH30994337 Life Chemicals Inc.

4-(pyrrolidin-2-yl)phenol hydrochloride

  • Cas Number : 7167-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart