LIFH3099433B Life Chemicals Inc.

N,1-dimethyl-1H-indazole-3-carboxamide

  • MDL number MFCD24556772
  • Cas Number : 335030-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart