LIFH3099433D Life Chemicals Inc.

2-[3-(fluoromethyl)azetidin-1-yl]ethan-1-ol hydrochloride

  • MDL number MFCD31626404
  • Cas Number : 2303565-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart