LIFH3099433E Life Chemicals Inc.

ethyl 4-(phenylsulfanyl)butanoate

  • MDL number MFCD11556293
  • Cas Number : 29193-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart