LIFH31555C88 Life Chemicals Inc.

1-benzyl-3-fluoropyrrolidine-3-carboxylic acid hydrochloride

  • Cas Number : 2307552-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart