LIFH31555C8A Life Chemicals Inc.

5-bromo-3-iodo-N,N,4-trimethylpyridin-2-amine

  • Cas Number : 2307553-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart