LIFH31555C9C Life Chemicals Inc.

methyl 6-(propan-2-yl)pyrimidine-4-carboxylate

  • MDL number MFCD30481860
  • Cas Number : 1616801-66-1
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-isopropylpyrimidine-4-carboxylate
Inchi Code 1S/C9H12N2O2/c1-6(2)7-4-8(9(12)13-3)11-5-10-7/h4-6H,1-3H3
Inchi Key DRKVALVACDWNRS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart