LIFH31555CA0 Life Chemicals Inc.

1-(4-(tert-butyl)cyclohexyl)azetidine-3-carboxylic acid

  • MDL number MFCD20954809
  • Cas Number : 1127402-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart