LIFH31555CA4 Life Chemicals Inc.

TERT-BUTYL 3-OXOPROPANOATE

  • MDL number MFCD22200887
  • Cas Number : 108350-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart