LIFH31555CB6 Life Chemicals Inc.

cyclooctane-1,3-dione

  • MDL number MFCD17015333
  • Cas Number : 935-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart