LIFH31555CB7 Life Chemicals Inc.

3-{hexahydro-1H-furo[3,4-c]pyrrole-5-carbonyl}piperidine hydrochloride

  • Cas Number : 2307552-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart