LIFH31555CB8 Life Chemicals Inc.

4,4-difluoro-1,4'-bipiperidine dihydrochloride

  • MDL number MFCD22566303
  • Cas Number : 1364631-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart