LIFH31555CBA Life Chemicals Inc.

2-(2-aminoethyl)isoindole-1,3-dione hydrochloride

  • MDL number MFCD08272836
  • Cas Number : 30250-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart