LIFH31555CBC Life Chemicals Inc.

tert-butyl N-(but-3-en-1-yl)-N-methylcarbamate

  • MDL number MFCD21648472
  • Cas Number : 312728-28-2
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-butenyl(methyl)carbamate
Inchi Code 1S/C10H19NO2/c1-6-7-8-11(5)9(12)13-10(2,3)4/h6H,1,7-8H2,2-5H3
Inchi Key MZYNQTOPYXEUDX-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart