LIFH31555CC5 Life Chemicals Inc.

3-(difluoromethyl)-2,6-difluorobenzaldehyde

  • MDL number MFCD28788409
  • Cas Number : 1806277-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart