LIFH31555CC6 Life Chemicals Inc.

6-bromo-3-(difluoromethyl)-2-fluorobenzaldehyde

  • MDL number MFCD31920075
  • Cas Number : 2092861-81-7
  • Molecular Weight : 253.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-(difluoromethyl)-2-fluorobenzaldehyde
Inchi Code 1S/C8H4BrF3O/c9-6-2-1-4(8(11)12)7(10)5(6)3-13/h1-3,8H
Inchi Key NIYWELFYYPPGDU-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart