LIFH31555CC7 Life Chemicals Inc.

2-bromo-3-(difluoromethyl)-6-fluorobenzaldehyde

  • Cas Number : 2092769-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart