LIFH31555CC8 Life Chemicals Inc.

4-(difluoromethyl)-2,3,6-trifluorobenzaldehyde

  • Cas Number : 2307552-74-3
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(difluoromethyl)-2,3,6-trifluorobenzaldehyde
Inchi Code 1S/C8H3F5O/c9-5-1-3(8(12)13)6(10)7(11)4(5)2-14/h1-2,8H
Inchi Key MJEOITGJLAYXQL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart