LIFH31555CCF Life Chemicals Inc.

3-methyl-1-[(piperidin-4-yl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride

  • Cas Number : 2307552-75-4
  • Molecular Weight : 259.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-1-(piperidin-4-ylmethyl)pyrimidine-2,4(1H,3H)-dione hydrochloride
Inchi Code 1S/C11H17N3O2.ClH/c1-13-10(15)4-7-14(11(13)16)8-9-2-5-12-6-3-9;/h4,7,9,12H,2-3,5-6,8H2,1H3;1H
Inchi Key ACYYKIGJBWVATD-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart