LIFH31555CD0 Life Chemicals Inc.

1-(piperidin-3-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride

  • Cas Number : 2307552-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart