LIFH324A10A3 Life Chemicals Inc.

1-hydroxycyclobutane-1-carboxylic acid

  • MDL number MFCD08861856
  • Cas Number : 41248-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart