LIFH324A10B1 Life Chemicals Inc.

2-(2-methoxyphenyl)thiazolidine

  • MDL number MFCD00805017
  • Cas Number : 40790-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart