LIFH324A10C8 Life Chemicals Inc.

4-(4-methoxyphenyl)-1,3-dimethyl-1H-pyrazol-5-amine

  • MDL number MFCD09055260
  • Cas Number : 1015846-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart